Anti-ITGB4 Mouse Monoclonal Antibody [clone: 450-9D]
Catalog # ABNOMAB5239
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 4
- Antigen symbol:ITGB4
- Clonality:Monoclonal
- Clone:450-9D
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3691
- Antigen synonyms:integrin subunit beta 4|AA407042|GP150|CD104
- Storage buffer:PBS, pH 7,4 (0,09% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human ITGB4.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against native ITGB4.
Recommended Dilutions: Flow Cytometry: 10?l of the s?ggested working dilution (1:25-1:100 to label 10^6 cells in 100?l; Immunohistochemistry (Frozen sections): 1:500-1:1000
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: 450-9D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human