Anti-LAAM(p) Polyclonal Antibody
Catalog # ABNOPAB8290
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:LAAM(p)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Cross adsorption:No
- Format:Liquid
- Storage buffer:20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:LAAM(p) conjugated with BSA.
- Pk:1 mg
Specifications
About this item
Sheep polyclonal antibody raised against LAAM(p).
Recommended Dilutions: Enzyme Immunoassay (0.25 ?g/ml
Type: Primary
Antigen: LAAM(p)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: