Order Entry
Sweden
ContactUsLinkComponent
Anti-TRIM5 Goat Polyclonal Antibody
Anti-TRIM5 Goat Polyclonal Antibody
Catalog # ABNOPAB6946
Supplier:  Abnova
Anti-TRIM5 Goat Polyclonal Antibody
Catalog # ABNOPAB6946
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tripartite Motif Containing 5
  • Antigen symbol:
    TRIM5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    85363
  • Antigen synonyms:
    TRIM5alpha|Tripartite motif-containing protein 5|RNF88|RING finger protein 88|TRIM5 alpha
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    CNIEKNENYQPK
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human TRIM5.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Goat polyclonal antibody raised against synthetic peptide of TRIM5.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: TRIM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: