Anti-CELSR3 Rabbit Polyclonal Antibody
ABNOPAB26117
: Abnova
- Antibody type:Primary
- Antigen name:Cadherin, EGF LAG seven-pass G-type receptor 3
- Antigen symbol:CELSR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hamster,Rabbit,Horse,Monkey,Gorilla,Human,Bovine,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:1951
- Antigen synonyms:RESDA1|FMI1|HFMI1|ADGRC3|MEGF2|CDHF11|EGFL1
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at N-terminus of human CELSR3.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of CELSR3.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-11 ?g/ml
Type: Primary
Antigen: CELSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat