Order Entry
Sweden
ContactUsLinkComponent
Anti-OR2A12 Rabbit Polyclonal Antibody
Anti-OR2A12 Rabbit Polyclonal Antibody
Catalog # ABGEAP19569B
Leverantör:  Abgent
Anti-OR2A12 Rabbit Polyclonal Antibody
Catalog # ABGEAP19569B
Leverantör:  Abgent

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    olfactory receptor, family 2, subfamily A, member 12
  • Antigen symbol:
    OR2A12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    346525
  • Antigen synonyms:
    OR2A12P|OR2A16P
  • Amino acid number:
    243 to 271
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    35.213 kDa
  • Sequence:
    YMAPKSSHSQERRKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.92
  • Shipping temperature:
    4 °C
  • Immunogen:
    This OR2A12 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human OR2A12.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifikationer

Om denna produkt

Type: Primary
Antigen: OR2A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human