Order Entry
Sweden
ContactUsLinkComponent
Anti-CRLF3 Rabbit Polyclonal Antibody
Anti-CRLF3 Rabbit Polyclonal Antibody
Catalog # ABGEAP17520B
Supplier:  Abgent
Anti-CRLF3 Rabbit Polyclonal Antibody
Catalog # ABGEAP17520B
Supplier:  Abgent

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    cytokine receptor-like factor 3
  • Antigensymbol:
    CRLF3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flödescytometri:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Gen-ID:
    51379
  • Antigen synonymer:
    CREME-9|CYTOR4|p48.2|CRLM9|CREME9|FRWS
  • Aminosyretal:
    305 to 331
  • Storage buffer:
    pH 7,4
  • Molekylvikt:
    49.766 kDa
  • Sequence:
    RVETVGQPDRRDC
  • Förvaringstemperatur:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Koncentration:
    0.7
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CRLF3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 305-331 amino acids from the C-terminal region of human CRLF3.
  • Rening:
    Protein A & Peptide affinity purification
  • Förp.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CRLF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human