Order Entry
Sweden
ContactUsLinkComponent
Anti-G2E3 Rabbit Polyclonal Antibody
Anti-G2E3 Rabbit Polyclonal Antibody
Catalog # ABGEAP18914C
Supplier:  Abgent
Anti-G2E3 Rabbit Polyclonal Antibody
Catalog # ABGEAP18914C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G2/M-phase specific E3 ubiquitin Protein ligase
  • Antigen symbol:
    G2E3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    55632
  • Antigen synonyms:
    KIAA1333|PHF7B
  • Amino acid number:
    226 to 252
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    80.504 kDa
  • Sequence:
    KEGRDYNAPDSKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.51
  • Shipping temperature:
    4 °C
  • Immunogen:
    This G2E3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 226-252 amino acids from the Central region of human G2E3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: G2E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human