Order Entry
Sweden
ContactUsLinkComponent
Anti-CASK Rabbit Polyclonal Antibody
Anti-CASK Rabbit Polyclonal Antibody
Catalog # ABGEAP20487C
Supplier:  Abgent
Anti-CASK Rabbit Polyclonal Antibody
Catalog # ABGEAP20487C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    calcium/calmodulin-dependent serine protein kinase (MAGUK family)
  • Antigen symbol:
    CASK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    29647
  • Amino acid number:
    589 to 616
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    103.259 kDa
  • Sequence:
    YVRAQFEYDPAKDC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.26
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Rat Cask antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 589-616 amino acids from the Central region of rat Cask.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CASK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Rat