Order Entry
Sweden
ContactUsLinkComponent
Anti-TIMM50 Rabbit Polyclonal Antibody
Anti-TIMM50 Rabbit Polyclonal Antibody
Catalog # ABGEAP19064A
Supplier:  Abgent
Anti-TIMM50 Rabbit Polyclonal Antibody
Catalog # ABGEAP19064A
Supplier:  Abgent

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae)
  • Antigensymbol:
    TIMM50
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Gen-ID:
    92609
  • Antigen synonymer:
    TIM50L|TIM50
  • Aminosyretal:
    74 to 104
  • Storage buffer:
    pH 7,4
  • Molekylvikt:
    39.646 kDa
  • Sequence:
    NPVDENGAKIPDEFDNC
  • Förvaringstemperatur:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Koncentration:
    0.57
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TIMM50 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 74-104 amino acids from the N-terminal region of human TIMM50.
  • Rening:
    Protein A & Peptide affinity purification
  • Förp.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TIMM50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human