Anti-CEBPD Rabbit Polyclonal Antibody
Catalog # ABGEAP20492C
Leverantör: Abgent
Specifikationer
- Antibody type:Primary
- Antigen name:CCAAT/enhancer binding protein (C/EBP), delta
- Antigen symbol:CEBPD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Gene ID:25695
- Antigen synonyms:C/EBPd|CELF
- Amino acid number:161 to 189
- Storage buffer:pH 7,4
- Molecular weight:28.6 kDa
- Sequence:PVREKGAGKRGPDRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This Rat Cebpd antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 161-189 amino acids from the Central region of rat Cebpd.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: CEBPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat