Order Entry
Sweden
ContactUsLinkComponent
Anti-CGREF1 Rabbit Polyclonal Antibody
Anti-CGREF1 Rabbit Polyclonal Antibody
Catalog # ABGEAP20491B
Leverantör:  Abgent
Anti-CGREF1 Rabbit Polyclonal Antibody
Catalog # ABGEAP20491B
Leverantör:  Abgent

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    Cell growth regulator with EF-Hand domain 1)
  • Antigen symbol:
    CGREF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    245918
  • Antigen synonyms:
    Cgr11
  • Amino acid number:
    220 to 248
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    30.835 kDa
  • Sequence:
    PREDAERQVESKDNC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This RAT Cgref1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 220-248 amino acids from the C-terminal region of rat Cgref1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifikationer

Om denna produkt

Type: Primary
Antigen: CGREF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Rat