Order Entry
Sweden
ContactUsLinkComponent
Anti-YWHAZ Mouse Monoclonal Antibody [clone: 1314CT423.108.153.173.140]
Anti-YWHAZ Mouse Monoclonal Antibody [clone: 1314CT423.108.153.173.140]
Catalog # ABGEAM2256A
Supplier:  Abgent
undefined
Anti-YWHAZ Mouse Monoclonal Antibody [clone: 1314CT423.108.153.173.140]
Catalog # ABGEAM2256A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta
  • Antigen symbol:
    YWHAZ
  • Clonality:
    Monoclonal
  • Clone:
    1314CT423.108.153.173.140
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    7534
  • Antigen synonyms:
    KCIP-1|HEL4|YWHAD|14-3-3-zeta|HEL-S-3
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    27.745 kDa
  • Sequence:
    MDKNELVQKAKLAEQAERYDDMAACMKSVTEQGAELSNEERNLLSVAYKNVVGARRSSWR
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.44
  • Shipping temperature:
    4 °C
  • Immunogen:
    This YWHAZ antibody is generated from a mouse immunized with a recombinant protein from human YWHAZ.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: YWHAZ
Clonality: Monoclonal
Clone: 1314CT423.108.153.173.140
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat