Order Entry
Sweden
ContactUsLinkComponent
Anti-TFAP2A Rabbit Polyclonal Antibody
Anti-TFAP2A Rabbit Polyclonal Antibody
Catalog # ABGEAP20558C
Supplier:  Abgent
Anti-TFAP2A Rabbit Polyclonal Antibody
Catalog # ABGEAP20558C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    transcription factor AP-2, alpha
  • Antigen symbol:
    TFAP2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    21418
  • Antigen synonyms:
    Ap2tf|Ap-2 (a)|AP2alpha|AP-2|Ap2|Tcfap2a
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    47.971 kDa
  • Sequence:
    HAIEDVPHVEDPG
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.7
  • Shipping temperature:
    4 °C
  • Immunogen:
    This mouse Tfap2a antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 134-166 amino acids from the Central region of mouse Tfap2a.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TFAP2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Mouse