Anti-PRLR Rabbit Polyclonal Antibody
Catalog # ABGEAP20518C
Leverantör: Abgent
Specifikationer
- Antibody type:Primary
- Antigen name:prolactin receptor
- Antigen symbol:PRLR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:5618
- Antigen synonyms:hPRLrI|MFAB|HPRL
- Amino acid number:147 to 179
- Storage buffer:pH 7,4
- Molecular weight:69.506 kDa
- Sequence:LYEIRLKPEKAAEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This PRLR antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 147-179 amino acids from the Central region of human PRLR.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: PRLR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human