Order Entry
Sweden
ContactUsLinkComponent
Anti-FABP4 Mouse Monoclonal Antibody [clone: 1105CT1.1.1]
Anti-FABP4 Mouse Monoclonal Antibody [clone: 1105CT1.1.1]
Catalog # ABGEAM2235B
Supplier:  Abgent
Anti-FABP4 Mouse Monoclonal Antibody [clone: 1105CT1.1.1]
Catalog # ABGEAM2235B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fatty Acid Binding Protein 4
  • Antigen symbol:
    FABP4
  • Clonality:
    Monoclonal
  • Clone:
    1105CT1.1.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    2167
  • Antigen synonyms:
    ALBP|A-FABP|AFABP|aP2|HEL-S-104
  • Amino acid number:
    1 to 132
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    14.719 kDa
  • Sequence:
    MCDAFVGTWKLVSSENFDDYMKEVGVGFATRKVAGMAKPNMIISVNGDVITIKSESTFKNTEISFILGQEFDEVTADDRKVKSTITLDGGVLVHVQKWDGKSTTIKRKREDDKLVVECVMKGVTSTRVYERA
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.48
  • Shipping temperature:
    4 °C
  • Immunogen:
    Purified His-tagged FABP4 protein was used to produced this monoclonal antibody.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: FABP4
Clonality: Monoclonal
Clone: 1105CT1.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse