Order Entry
Sweden
ContactUsLinkComponent
Anti-FYN Mouse Monoclonal Antibody [clone: 1302CT390.118.237]
Anti-FYN Mouse Monoclonal Antibody [clone: 1302CT390.118.237]
Catalog # ABGEAM2266B
Supplier:  Abgent
undefined
Anti-FYN Mouse Monoclonal Antibody [clone: 1302CT390.118.237]
Catalog # ABGEAM2266B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FYN proto-oncogene, Src family tyrosine kinase
  • Antigen symbol:
    FYN
  • Clonality:
    Monoclonal
  • Clone:
    1302CT390.118.237
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    2534
  • Antigen synonyms:
    SLK|SYN|p59-FYN
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    60.762 kDa
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    2.000
  • Shipping temperature:
    4 °C
  • Immunogen:
    This FYN antibody is generated from a mouse immunized with a recombinant preotein from human FYN.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: FYN
Clonality: Monoclonal
Clone: 1302CT390.118.237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human