Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IFNA4 Rabbit Polyclonal Antibody
Catalog # ABGEAP20476B
Supplier:  Abgent
Anti-IFNA4 Rabbit Polyclonal Antibody
Catalog # ABGEAP20476B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon alpha 4
  • Antigen symbol:
    IFNA4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    3441
  • Antigen synonyms:
    IFN-alpha4a|INFA4
  • Amino acid number:
    162 to 189
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    21.808 kDa
  • Sequence:
    CFSTNLQKRLRRKD
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.7
  • Shipping temperature:
    4 °C
  • Immunogen:
    This IFNA4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 162-189 amino acids from the C-terminal region of human IFNA4.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: IFNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human