Order Entry
Sweden
ContactUsLinkComponent
Anti-SSNA1 Rabbit Polyclonal Antibody
Anti-SSNA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP20483A
Leverantör:  Abgent
Anti-SSNA1 Rabbit Polyclonal Antibody
Catalog # ABGEAP20483A
Leverantör:  Abgent

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    Sjogren syndrome nuclear autoantigen 1
  • Antigen symbol:
    SSNA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    8636
  • Antigen synonyms:
    N14|NA14|NA-14
  • Amino acid number:
    25 to 53
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    13.596 kDa
  • Sequence:
    KQRLQNEVRQLTEKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This SSNA1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 25-53 amino acids from the N-terminal region of human SSNA1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifikationer

Om denna produkt

Type: Primary
Antigen: SSNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human