Order Entry
Sweden
ContactUsLinkComponent
Anti-PAF Mouse Monoclonal Antibody [clone: 11A4]
Anti-PAF Mouse Monoclonal Antibody [clone: 11A4]
Catalog # CAYM160600-1
Leverantör:  Cayman Chemical
Anti-PAF Mouse Monoclonal Antibody [clone: 11A4]
Catalog # CAYM160600-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antibody type:
    Primary
  • Antigen symbol:
    PAF
  • Clonality:
    Monoclonal
  • Clone:
    11A4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Human,
    Bovine
  • Cross adsorption:
    No
  • Amino acid number:
    260 to 269
  • Storage buffer:
    TBS, pH 7,4
  • Sequence:
    LGFQDSKFHQ
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human PAF receptor amino acids 260-269
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: PAF
Clonality: Monoclonal
Clone: 11A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Pig