Anti-Zrp Mouse Monoclonal Antibody [clone: MN-8C4]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Zrp
Clonality: Monoclonal
Clone: MN-8C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Arctic Char, Atlantic Salmon, Cod, Rainbow Trout, Turbot
Specifications
- Catalog No:
- CAYM100017-100
- CAYM100017-500
- Antigen symbol:
- Zrp
- Zrp
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- MN-8C4
- MN-8C4
- Reactivity:
- Arctic char, Rainbow trout, Cod, Turbot, Atlantic salmon
- Arctic char, Rainbow trout, Cod, Turbot, Atlantic salmon
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- eggshell proteins (zona radiata proteins and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
- eggshell proteins (zona radiata proteins and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C