Anti-PPI Goat Polyclonal Antibody
Catalog # ROCK200-1174S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:PPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Carica papaya
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Papain Antibody was produced by repeated immunizations with pork Papain protein isolated from papaya.
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:25 µl
- Pk:25 µl
Specifications
About this item
Anti-Papain antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Researchers should determine optimal titers for applications that are not stated below.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:3000; Other Dilution: User Optimized
Type: Primary
Antigen: PPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Carica papaya