Anti-HRPC Guinea Pig Polyclonal Antibody
Catalog # ROCK200-2138S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen symbol:PRXC, HRPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Guinea pig
- ImmunoChemistry:Yes
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Peroxidase [Horseradish]
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:25 µl
- Pk:25 µl
Specifications
About this item
Anti-Peroxidase (Horseradish) (Guinea Pig) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized
Type: Tag
Antigen: HRPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: