Order Entry
Sweden
ContactUsLinkComponent
Anti-AKT1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14E5.A2.B2.H9]
Anti-AKT1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14E5.A2.B2.H9]
Catalog # ROCK200-301-I51S
Anti-AKT1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14E5.A2.B2.H9]
Catalog # ROCK200-301-I51S

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase B alpha
  • Antigen symbol:
    PKBA
  • Clonality:
    Monoclonal
  • Clone:
    14E5.A2.B2.H9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    No
  • Antigen synonyms:
    AKT
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-AKT1 Antibody was produced by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 protein.
  • Purification:
    Anti-AKT1 antibody is directed against human AKT1. The antibody detects both unphosphorylated and phosphorylated forms of the protein. Anti-AKT1 antibody was purified from ascites by Protein A chromatography. Cross reactivity with AKT1 from other species has not been determined, however, the sequence of the immunogen shows 85% identity to mouse and 92% identity with rat, therefore, cross reactivity is expected.
  • Size:
    25 µl
  • Pk:
    25 µl

Specifications

About this item

Anti-AKT1 Antibody is suitable for ELISA, immunohistochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Flow Cytometry: User Optimized; Western Blot: 1:1000; Immunochemistry: 20 µg/ml; Other Dilution: User Optimized

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human