Anti-ORM1 Rabbit Polyclonal Antibody
ROCK200-401-046S
- Antibody type:Primary
- Antigen name:orosomucoid 1
- Antigen symbol:ORM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5004
- Antigen synonyms:AGP1|HEL-S-153w|ORM|AGP-A
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Alpha-1-Acid Glycoprotein Antibody was produced by repeated immunizations with human plasma a-1-Acid Glycoprotein.
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:25 µl
- Pk:25 µl
Anti-Alpha-1-Acid Glycoprotein Antibody is suitable for western blottingand for ELISA. Researchers should determine optimal titers for applications that are not stated below.
Recommended Dilutions: ELISA: 1:135000; Western Blot: 1:2000 - 1:10000; Immunochemistry: User Optimized; Other Dilution: User Optimized
Type: Primary
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human