Anti-HRPC Sheep Polyclonal Antibody
Catalog # ROCK200-6138-0100
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen symbol:PRXC, HRPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Peroxidase [Horseradish]
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:3000; Other Dilution: User Optimized
Type: Tag
Antigen: HRPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: