Anti-MDH1 Sheep Polyclonal Antibody
Catalog # ROCK200-601-145S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:malate dehydrogenase 1, NAD (soluble)
- Antigen symbol:MDH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Reactivity:Pig
- Western blot:Yes
- Cross adsorption:No
- Gene ID:396894
- Antigen synonyms:MDH
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Malate Dehydrogenase [Pig Heart]
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:25 µl
- Pk:25 µl
Specifications
About this item
MALATE DEHYDROGENASE antibody has been assayed against 1.0 ug of Malate Dehydrogenase [Pig Heart] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100; Other Dilution: User Optimized
Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Pig