Anti-AHSP Mouse Polyclonal Antibody
Catalog # ROCK100-401-E79S
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Alpha hemoglobin stabilizing protein
- Antigensymbol:AHSP
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- ImmunKemi:Yes
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Gen-ID:51327
- Antigen synonymer:ERAF|EDRF
- Storlek:25 µl
- Förp.:25 µl
Specifications
About this item
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse