Anti-GFP Mouse Monoclonal Antibody (DyLight® 649) [clone: 9F9.F9]
Katalog # ROCK200-343-215
Leverantör: Rockland Immunochemicals
Specifikationer
- Antibody type:Tag
- Antigen symbol:GFP
- Clonality:Monoclonal
- Clone:9F9.F9
- Conjugation:DyLight® 649
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Multiple
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BALB/c mice were immunized with a Green Fluorescent Protein (GFP) fusion Protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria. A hybridoma was produced by the fusion of BALB/c mouse splenocyte
- Tested applications:Fluoresence ELISA
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifikationer
Om denna produkt
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: ELISA: 1:20000-1:40000; FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000-1:25000;
Restore with deionized water (or equivalent); 100 µl
Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: DyLight® 649
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan