Anti-GDF15 Rabbit Polyclonal Antibody
ROCK610-401-B87
- Antibody type:Primary
- Antigen name:Growth differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- ChIP:Yes
- Gene ID:23886
- Antigen synonyms:MIC-1|SBF|NAG-1
- Size:100 µg
- Pk:100 µG
This affinity purified antibody is suitable for ELISA and western blotting of mouse and human NAG-1 Protein.
Recommended Dilutions: ELISA: 1:100000 - 1:120000; Western Blot: 1:3000 - 1:7000;
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human