Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 800)
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 800)
Catalog # ROCK605-745-002-05
undefined
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 800)
Catalog # ROCK605-745-002-05

Specifikationer

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 800
  • Host:
    Donkey
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Size:
    500 µg
  • Pk:
    500 µG

Specifikationer

Om denna produkt

Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat