Anti-HIST2H3C Rabbit Polyclonal Antibody
Katalog # ROCK600-401-I69
Leverantör: Rockland Immunochemicals
Specifikationer
- Antibody type:Primary
- Antigen name:histone cluster 2, H3c
- Antigen symbol:HIST2H3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Drosophila,Plant,Xenopus,Chicken,C. elegans,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:126961
- Antigen synonyms:H3F2|H3.2|H3FM|H3|H3/M|H3FN
- Modification:Monometyl-Lys9
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Histone H3 [Monometyl-Lys9] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic monomethylated peptide surrounding Lysine 9 of human Histone H3.2.
- Tested applications:Dot Blot
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifikationer
Om denna produkt
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:500; Western Blot: 1:500; Immunochemistry: 1:500; Other Dilution: Dot Blot 0.5µg/ml
Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant