Anti-BD1 Rabbit Polyclonal Antibody
Catalog # USBIB0899-03K-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:BD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Amino acid number:1 to 5
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human β-Defensin 1 (aa 1-5) polylysinee conjugated (DHYNC(ACM)GG)
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BD1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,