Anti-SNAP Tag Rabbit Polyclonal Antibody
USBIS1014-67Y-40
- Antibody type:Primary
- Antigen symbol:SNAP Tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Purified recombinant SNAP-tag fusion protein
- Purification:purified by immunoaffinity chromatography
- Pk:40 µG