Anti-Phosphotyrosine Mouse Monoclonal Antibody [clone: 6D669]
Catalog # USBIP4078-37-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Phosphotyrosine
- Antigen symbol:pY
- Clonality:Monoclonal
- Clone:6D669
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Immunogen:Phosphotyrosine containing peptides.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Phosphotyrosine
Clonality: Monoclonal
Clone: 6D669
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat,