Anti-Histone Nucleosome Core Sheep Polyclonal Antibody
Catalog # USBIH5110-01C-250
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Histone Nucleosome Core
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Bovine
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, before the addition of glycerolto 40%.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Intact calf histone nucleosome core complexed to rRNA. Molecular Weight: 15kD.
- Purification:purified by Protein G chromatography
- Size:250 μg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: Histone Nucleosome Core
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine,