Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 800)
Catalog # AGRIAS122393
Leverantör: Agrisera
Specifikationer
- Antibody type:Secondary
- Antigen symbol:IgE epsilon
- Clonality:Polyclonal
- Conjugation:DyLight® 800
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified Human IgE, (epsilon) chain
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifikationer
Om denna produkt
Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype:
Reactivity: Human