Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 800)
Catalog # AGRIAS122379
Supplier: Agrisera
Specifications
- Antikroppstyp:Sekundär
- Antigensymbol:IgG (H+L)
- Klonalitet:Polyclonal
- Konjugation:DyLight® 800
- Flödescytometri:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Goat
- Western blot:Yes
- Korsad adsorption:No
- Förvaringstemperatur:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified Goat IgG, whole molecule
- Rening:purified
- Storlek:1 mg
- Förp.:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat