Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 650)
Catalog # AGRIAS122332
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified Rabbit IgG, whole molecule
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit