Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody
Anti-IgG F(c) Goat Polyclonal Antibody
Catalog # AGRIAS10799
Leverantör:  Agrisera
Anti-IgG F(c) Goat Polyclonal Antibody
Catalog # AGRIAS10799
Leverantör:  Agrisera

Specifikationer

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG F(c)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Reactivity:
    Rabbit,
    Human,
    Bovine,
    Mouse
  • Seq:
    01
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    purified human IgG,purified human IgG
  • Purification:
    affinity-purified
  • Size:
    2 mg
  • Pk:
    2 mg

Specifikationer

Om denna produkt

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit