Anti-IgG (H+L) Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # AGRIAS101167
Leverantör: Agrisera
Specifikationer
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Chicken
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Purified Mouse IgG, whole molecule
- Tested applications:Dot Blot
- Purification:purified
- Size:500 μg
- Pk:0,5 mg
Specifikationer
Om denna produkt
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse