Anti-IgG (H+L) Goat Polyclonal Antibody
AGRIAS101108
: Agrisera
- Antikroppstyp:Sekundär
- Antigensymbol:IgG (H+L)
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Goat
- Reaktivitet:Goat,Human
- Korsad adsorption:No
- Förvaringstemperatur:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified Rat IgG, whole molecule
- Rening:affinity-purified
- Storlek:1 mg
- Förp.:1 mg
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Goat, Human