Anti-Soy protein Chicken Polyclonal Antibody
Catalog # AGRIAS09569
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:Soy Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:+4 °C
- Shipping temperature:Ambient
- Immunogen:Glycine max flour protein extract
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Soy Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: