Anti-PsbS Chicken Polyclonal Antibody
Catalog # AGRIAS03032
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PSBS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Zea mays,Pisum sativum,A. thaliana,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Molecular weight:22 kDa
- Storage temperature:+4 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from available di and monocot PsbS sequences, including Arabidopsis thaliana (At1g44575). This sequence is even conserved in conifers.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PSBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: A. thaliana, H. vulgare, Pisum sativum, Zea mays