Anti-GlnA Chicken Polyclonal Antibody
Catalog # AGRIAS01018
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:GLNA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Spinacia oleracea,Gracilaria gracilis,Red algae,Zea mays,A. thaliana,Eragrostis tef,Physcomitrella patens
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:GLN2
- Storage temperature:+4 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from available bacterial GlnA sequences with perfect conservation in alpha, beta, gamma Proteobacteria, Enterobacteria, Thermotogales, Low GC Gram+, Cyanobacteria (except weak conservation with Trichodesmium thiebautii) including Synechocystis PCC 6803 Q59981
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GLNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: A. thaliana, Eragrostis tef, Physcomitrella patens, Spinacia oleracea, Zea mays, Red Algae, Gracilaria gracilis