Anti-Lhca3 Rabbit Polyclonal Antibody
Catalog # AGRIAS01007
Leverantör: Agrisera
Specifikationer
- Antibody type:Primary
- Antigen symbol:LHCA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Phaseolus vulgaris,Arachis hypogaea,Physcomitrella patens,Oryza sativa,Spinacia oleracea,Zea mays,Guzmania hybrid,Pisum sativum,A. thaliana,N. tabacum,Solanum lycopersicum,Pinus strobus,Hordeum vulgare,C. quitensis kunt bartl
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9SY97
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated synthetic peptide derived from the Lhca3 protein sequence from Arabidopsis thaliana
- Size:200 µl
- Pk:200 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: LHCA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Arachis hypogaea, Colobanthus quitensis kunt bartl, Guzmania hybrid, Hordeum vulgare, Solanum lycopersicum, N. tabacum, Oryza sativa, Phaseolus vulgaris, Physcomitrella patens, Pinus strobus, Pisum sativum, Spinacia oleracea, Zea mays