Anti-Lhcb3 Rabbit Polyclonal Antibody
AGRIAS01002
: Agrisera
- Antibody type:Primary
- Antigen symbol:LHCB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:C. quitensis kunt bartl,Phaseolus vulgaris,Arachis hypogaea,Physcomitrella patens,Oryza sativa,Spinacia oleracea,Miscanthus giganteus,C. reinhardtii,Mesembryanthemum crystallinum,Zea mays,Pisum sativum,A. thaliana,N. tabacum,Solanum lycopersicum,Hordeum vulgare
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9S7M0
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated synthetic peptide derived from a highly conserved sequence of Lhcb3 proteins from angiosperms (monocots and dicots) and gymnosperms, including Arabidopsis thaliana Lhcb3
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: LHCB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Arachis hypogaea, Colobanthus quitensis kunt bartl, Hordeum vulgare, Solanum lycopersicum, Mesembryanthemum crystallinum, Miscanthus giganteus, N. tabacum, Oryza sativa, Phaseolus vulgaris, Physcomitrella patens, Pisum sativum, Spinacia oleracea, Zea mays, C. reinhardtii