Anti-ClpC Rabbit Polyclonal Antibody
Catalog # AGRIAS01001
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:CLPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Synechococcus sp.,C. reinhardtii,A. thaliana,Streptomyces sp.
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant ClpC (C-terminal domain overexpressed as fusion with maltose-binding protein) Q55023
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CLPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: A. thaliana, C. reinhardtii, Streptomyces sp., Synechococcus sp.