Anti-FVIIa Rabbit Polyclonal Antibody
Catalog # AGRIAS00001-500
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:FVIIA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Plant
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant human Factor VIIa (NovoSeven ®, Novo Nordisk A/S Denmark)
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: FVIIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: higher Plants