Anti-KRT20 Mouse Monoclonal Antibody [clone: UMAB167]
Katalog # ORIGUM870060
Leverantör: OriGene
Specifikationer
- Antibody type:Primary
- Antigen symbol:KRT20
- Clonality:Monoclonal
- Clone:UMAB167
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:48.3 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human KRT20 (NP_061883) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:30 µl
Specifikationer
Om denna produkt
Recommended Dilutions: Immunochemistry: 1:100-200
Type: Primary
Antigen: KRT20
Clonality: Monoclonal
Clone: UMAB167
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human