Anti-TYMP Mouse Monoclonal Antibody [clone: UMAB90]
Catalog # ORIGUM800021
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TYMP
- Clonality:Monoclonal
- Clone:UMAB90
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:311 to 482
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:49.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 311-482 of human TYMP (NP_001944) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100
Type: Primary
Antigen: TYMP
Clonality: Monoclonal
Clone: UMAB90
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human